Please login to your online account to display your discounted pricing

anti-FosB/G0S3, Clone: 83B1138, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056530

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056530

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Formulation PBS containing 0.05% BSA
Gene ID (Entrez) 2354
Conjugate Unlabeled
Test Specificity In HeLa, a 46 kDa band is observed.
Concentration 0.5mg/mL
Gene Accession No. P53539
Host Species Mouse
Quantity 0.1mg
Primary or Secondary Primary
Format Purified
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
Regulatory Status RUO
Clone 83B1138
Antigen FosB/G0S3
Applications Western Blotting
Isotype IgG1
Cross Reactivity Human