Please login to your online account to display your discounted pricing

anti-FITC, Clone: 2A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1107898SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1107898SS

  • / Each
 

Description & Specifications

Specifications

Antigen FITC
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone 2A3
Conjugate Unlabeled
Cross Reactivity Non-species specific
Format Purified
Formulation PBS. pH 7.4
Host Species Murine
Immunogen FITC-BSA conjugate.
Isotype IgG2a
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This product reacts with both free fluorescein (FITC) and conjugated FITC, so it recognizes FITC-labeled proteins. Kd = 1.5 x 10-8 M. Detects FITC labelled products.
Target Species Non-species Specific