Please login to your online account to display your discounted pricing

anti-FITC, Clone: 2A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1107898

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1107898

  • / Each
 

Description & Specifications

Specifications

Test Specificity This product reacts with both free fluorescein (FITC) and conjugated FITC, so it recognizes FITC-labeled proteins. Kd = 1.5 x 10-8 M. Detects FITC labelled products.
Quantity 0.5mg
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone 2A3
Immunogen FITC-BSA conjugate.
Antigen FITC
Formulation PBS. pH 7.4
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype IgG2a
Cross Reactivity Non-species specific
Regulatory Status RUO
Host Species Mouse