Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: TV-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328491ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328491M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Immunogen T-cell lymphoma biopsy
Antigen Fibronectin/Anastellin
Regulatory Status RUO
Isotype κ
Host Species Mouse
Clone TV-1
Gene Accession No. P02751
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Format Purified
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 2335
Conjugate Unlabeled
Quantity 1mL