Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: TV-1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232849

 View all options for this product


For Research Use Only

Catalog No. NBP232849


Description & Specifications


Antigen Fibronectin/Anastellin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone TV-1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02751
Host Species Mouse
Immunogen T-cell lymphoma biopsy
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335