Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: HFN7.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232847

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232847

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Fibronectin/Anastellin
Regulatory Status RUO
Format Purified
Clone HFN7.1
Host Species Mouse
Isotype κ
Cross Reactivity Human
Primary or Secondary Primary
Gene ID (Entrez) 2335
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene Accession No. P02751
Concentration 0.2mg/mL
Conjugate Unlabeled
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Quantity 0.5mL