Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: HFN7.1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HFN7.1
Host Species Mouse
Isotype κ
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Antigen Fibronectin/Anastellin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328471M

 
novus biologicals canada
NBP2328471ML
Unlabeled 1mL Each for $429.00

NBP232847

 
novus biologicals canada
NBP232847
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HFN7.1
Host Species Mouse
Isotype κ
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Antigen Fibronectin/Anastellin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P02751
Gene ID (Entrez) 2335
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL