Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: HFN7.1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328471ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328471M


Description & Specifications


Antigen Fibronectin/Anastellin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone HFN7.1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02751
Host Species Mouse
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335