Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222113

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222113

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 2335
Conjugate Unlabeled
Formulation PBS
Primary or Secondary Primary
Host Species Mouse
Immunogen Purified recombinant fragment of human Fibronectin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02751]
Quantity 0.1mL
Isotype IgG1
Regulatory Status RUO
Cross Reactivity Human
Format Precipitation
Antigen Fibronectin/Anastellin
Gene Accession No. P02751
Monoclonal or Polyclonal Monoclonal
Clone 2F4
Concentration 1.0mg/mL