Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2F4, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 2F4
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Fibronectin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02751]
Antigen Fibronectin/Anastellin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222113SS

 
novus biologicals canada
NBP222113SS
Unlabeled 0.025mL Each for $139.00

NBP222113

 
novus biologicals canada
NBP222113
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 2F4
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Fibronectin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02751]
Antigen Fibronectin/Anastellin
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P02751
Gene ID (Entrez) 2335
Formulation PBS
Concentration 1.0mg/mL