Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222113SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222113SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Monoclonal
Antigen Fibronectin/Anastellin
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Precipitation
Quantity 0.025mL
Regulatory Status RUO
Immunogen Purified recombinant fragment of human Fibronectin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02751]
Isotype IgG1
Host Species Mouse
Cross Reactivity Human
Clone 2F4
Conjugate Unlabeled
Concentration 1.0mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 2335
Gene Accession No. P02751