Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2755-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328511ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328511M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Clone 2755-8
Regulatory Status RUO
Conjugate Unlabeled
Gene Accession No. P02751
Primary or Secondary Primary
Gene ID (Entrez) 2335
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen T-cell lymphoma biopsy
Concentration 0.2mg/mL
Antigen Fibronectin/Anastellin
Host Species Mouse
Quantity 1mL