Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2755-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232851

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232851

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen T-cell lymphoma biopsy
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Regulatory Status RUO
Antigen Fibronectin/Anastellin
Isotype κ
Gene Accession No. P02751
Primary or Secondary Primary
Gene ID (Entrez) 2335
Concentration 0.2mg/mL
Format Purified
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Clone 2755-8
Conjugate Unlabeled
Quantity 0.5mL