Please login to your online account to display your discounted pricing

anti-Fibronectin/Anastellin, Clone: 2755-8, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 2755-8
Host Species Mouse
Isotype κ
Immunogen T-cell lymphoma biopsy
Antigen Fibronectin/Anastellin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232851

 
novus biologicals canada
NBP232851
Unlabeled 0.5mL Each for $321.00

NBP2328511M

 
novus biologicals canada
NBP2328511ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 2755-8
Host Species Mouse
Isotype κ
Immunogen T-cell lymphoma biopsy
Antigen Fibronectin/Anastellin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P02751
Gene ID (Entrez) 2335
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL