Please login to your online account to display your discounted pricing

anti-Fibrinogen, Clone: 40F11, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12010068SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12010068T

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Format Purified
Immunogen Fibrin degradation products.
Antigen Fibrinogen
Cross Reactivity Human
Test Specificity Fibrinogen, Fibrin degradation products.
Formulation PBS. pH 7.4
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2243
Conjugate Unlabeled
Primary or Secondary Primary
Isotype IgG1
Applications Sandwich ELISA
Applications Western Blotting
Host Species Mouse
Quantity 0.04mg
Clone 40F11