Please login to your online account to display your discounted pricing

anti-Fibrinogen, Clone: 40F11, Novus Biologicals™

$110.17 - $222.95

Specifications

Clone 40F11
Host Species Murine
Isotype IgG1
Immunogen Fibrin degradation products.
Antigen Fibrinogen
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12010068

 
novus biologicals canada
NB12010068
Unlabeled 0.2mg Each for $222.95

NB12010068T

 
novus biologicals canada
NB12010068SS
Unlabeled 0.04mg Each for $110.17
Description & Specifications

Specifications

Clone 40F11
Host Species Murine
Isotype IgG1
Immunogen Fibrin degradation products.
Antigen Fibrinogen
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Sandwich ELISA
Applications Western Blot
Test Specificity Fibrinogen, Fibrin degradation products.
Gene ID (Entrez) 2243
Formulation PBS. pH 7.4