Please login to your online account to display your discounted pricing

anti-FGF R1, Clone: M19B2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001287

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001287

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1
Format Purified
Quantity 0.1mL
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Formulation PBS
Gene ID (Entrez) 2260
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Reacts with both alpha and beta isoforms.
Gene Accession No. P11362
Immunogen Recombinant human ectodomain of FGFR1b expressed in &italic_begin;E. coli&italic_end; beginning with proline 23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
Clone M19B2
Host Species Mouse
Antigen FGF R1