Please login to your online account to display your discounted pricing

anti-FGF R1, Clone: M19B2, Novus Biologicals™

$144.63 - $362.00

Specifications

Clone M19B2
Host Species Murine
Isotype IgG1
Immunogen Recombinant human ectodomain of FGFR1b expressed in &italic_begin;E. coli&italic_end; beginning with proline 23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
Antigen FGF R1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001287SS

 
novus biologicals canada
NB6001287SS
Unlabeled 0.025mL Each for $144.63

NB6001287

 
novus biologicals canada
NB6001287
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Clone M19B2
Host Species Murine
Isotype IgG1
Immunogen Recombinant human ectodomain of FGFR1b expressed in &italic_begin;E. coli&italic_end; beginning with proline 23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
Antigen FGF R1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Reacts with both alpha and beta isoforms.
Gene Accession No. P11362
Gene ID (Entrez) 2260
Formulation PBS
Concentration 1.0mg/mL