Please login to your online account to display your discounted pricing

anti-FGF-23, Clone: FGF23/638, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328461ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328461M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Regulatory Status RUO
Quantity 1mL
Antigen FGF-23
Clone FGF23/638
Gene Accession No. Q9GZV9
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Conjugate Unlabeled
Concentration 0.2mg/mL
Gene ID (Entrez) 8074
Isotype κ
Format Purified
Applications ELISA
Immunogen Recombinant human FGF23 protein
Host Species Mouse