Please login to your online account to display your discounted pricing

anti-FGF-23, Clone: FGF23/638, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232846

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232846

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Clone FGF23/638
Isotype κ
Applications ELISA
Immunogen Recombinant human FGF23 protein
Host Species Mouse
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q9GZV9
Concentration 0.2mg/mL
Gene ID (Entrez) 8074
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Format Purified
Regulatory Status RUO
Antigen FGF-23