Please login to your online account to display your discounted pricing

anti-FGF-23, Clone: FGF23/638, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone FGF23/638
Host Species Mouse
Isotype κ
Immunogen Recombinant human FGF23 protein
Antigen FGF-23
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232846

 
novus biologicals canada
NBP232846
Unlabeled 0.5mL Each for $321.00

NBP2328461M

 
novus biologicals canada
NBP2328461ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone FGF23/638
Host Species Mouse
Isotype κ
Immunogen Recombinant human FGF23 protein
Antigen FGF-23
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. Q9GZV9
Gene ID (Entrez) 8074
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL