Please login to your online account to display your discounted pricing

anti-Fc gamma RIIIA/CD16a, Clone: 3G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP150056SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP150056SS

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Human neutrophils.
Format Purified
Antigen Fc gamma RIIIA/CD16a
Regulatory Status RUO
Quantity 0.02mg
Cross Reactivity Human
Cross Reactivity Primate
Clone 3G8
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal
Test Specificity The mouse monoclonal antibody 3G8 recognizes CD16, a low affinity receptor for aggregated IgG (FcgammaRIII antigen). CD16 exists in two different isoforms: CD16a (FcgammaRIIIA; 50-65 kDa; expressed on NK-cells, monocytes and macrophages) and CD16b (FcgammaRIIIB; 48 kDa; mainly expressed on neutrophils). HLDA V; WS Code NK80.
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS, pH 7.4
Gene ID (Entrez) 2214
Isotype IgG1