Please login to your online account to display your discounted pricing

anti-FANCD2, Clone: FI-17, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone FI-17
Host Species Murine
Immunogen Human FANCD2 fusion protein [Swiss-Prot No. Q9BXW9]
Antigen FANCD2
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100316SS

 
novus biologicals canada
NB100316SS
Unlabeled 0.025mL Each for $139.00

NB100316

 
novus biologicals canada
NB100316
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone FI-17
Host Species Murine
Immunogen Human FANCD2 fusion protein [Swiss-Prot No. Q9BXW9]
Antigen FANCD2
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Human. This antibody does not react with mouse or hamster.
Gene Accession No. Q9BXW9
Gene ID (Entrez) 2177