Please login to your online account to display your discounted pricing

anti-FANCC, Clone: 8F3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118974SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118974SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Format Purified
Gene Accession No. Q00597
Antigen FANCC
Host Species Mouse
Immunogen A synthetic peptide made to a region of the C-terminus of human FANCC (within residues 450-558) [Swiss-Prot No. Q00597].
Isotype IgG2b
Clone 8F3
Regulatory Status RUO
Gene ID (Entrez) 2176
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.9mg/mL
Conjugate Unlabeled