Please login to your online account to display your discounted pricing

anti-FANCC, Clone: 8F3, Novus Biologicals™

$139.00 - $321.00

Specifications

Clone 8F3
Host Species Murine
Isotype IgG2b
Immunogen A synthetic peptide made to a region of the C-terminus of human FANCC (within residues 450-558) [Swiss-Prot No. Q00597].
Antigen FANCC
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118974

 
novus biologicals canada
NBP118974
Unlabeled 0.1mL Each for $321.00

NBP118974SS

 
novus biologicals canada
NBP118974SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 8F3
Host Species Murine
Isotype IgG2b
Immunogen A synthetic peptide made to a region of the C-terminus of human FANCC (within residues 450-558) [Swiss-Prot No. Q00597].
Antigen FANCC
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q00597
Gene ID (Entrez) 2176
Formulation PBS
Concentration 1.9mg/mL