Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Clone: 5G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118973SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118973SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2175
Quantity 0.025mL
Format Purified
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Host Species Mouse
Immunogen A synthetic peptide made to a C-terminal region of human FANCA (within amino acids 1380-1455) [Swiss-Prot No. O15360].
Applications Immunoprecipitation
Applications Western Blotting
Antigen FACA/FANCA
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Clone 5G9
Conjugate Unlabeled
Gene Accession No. O15360
Concentration 2mg/mL
Formulation PBS