Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Clone: 5G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118973

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118973

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2175
Immunogen A synthetic peptide made to a C-terminal region of human FANCA (within amino acids 1380-1455) [Swiss-Prot No. O15360].
Isotype IgG1
Antigen FACA/FANCA
Regulatory Status RUO
Clone 5G9
Cross Reactivity Human
Format Purified
Host Species Mouse
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Formulation PBS
Gene Accession No. O15360
Primary or Secondary Primary
Concentration 2mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal