Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Clone: 5G9, Novus Biologicals™

$139.00 - $321.00

Specifications

Clone 5G9
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide made to a C-terminal region of human FANCA (within amino acids 1380-1455) [Swiss-Prot No. O15360].
Antigen FACA/FANCA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118973

 
novus biologicals canada
NBP118973
Unlabeled 0.1mL Each for $321.00

NBP118973SS

 
novus biologicals canada
NBP118973SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 5G9
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide made to a C-terminal region of human FANCA (within amino acids 1380-1455) [Swiss-Prot No. O15360].
Antigen FACA/FANCA
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. O15360
Gene ID (Entrez) 2175
Formulation PBS
Concentration 2mg/mL