Please login to your online account to display your discounted pricing

anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329661ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329661M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 1mL
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Regulatory Status RUO
Clone ER503
Antigen Estrogen Receptor
Gene Accession No. P03372
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2099
Concentration 0.2mg/mL
Conjugate Unlabeled
Isotype IgG1
Cross Reactivity Human