Please login to your online account to display your discounted pricing

anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232966

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232966

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Host Species Mouse
Isotype IgG1
Antigen Estrogen Receptor
Clone ER503
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 2099
Gene Accession No. P03372
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Format Purified