Please login to your online account to display your discounted pricing

anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone ER503
Host Species Mouse
Isotype IgG1
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Antigen Estrogen Receptor
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232966

 
novus biologicals canada
NBP232966
Unlabeled 0.5mL Each for $321.00

NBP2329661M

 
novus biologicals canada
NBP2329661ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone ER503
Host Species Mouse
Isotype IgG1
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Antigen Estrogen Receptor
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P03372
Gene ID (Entrez) 2099
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL