Please login to your online account to display your discounted pricing

anti-Erythrocyte Specific, Clone: SFL23.6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232845

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232845

  • / Each
 

Description & Specifications

Specifications

Antigen Erythrocyte Specific
Host Species Mouse
Quantity 0.5mL
Clone SFL23.6
Immunogen Hepatocytes from 20-22 week-old human fetus
Isotype κ
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Format Purified
Conjugate Unlabeled
Cross Reactivity Human