Please login to your online account to display your discounted pricing

anti-Erythrocyte Specific, Clone: SFL23.6, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone SFL23.6
Host Species Mouse
Isotype κ
Immunogen Hepatocytes from 20-22 week-old human fetus
Antigen Erythrocyte Specific
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232845

 
novus biologicals canada
NBP232845
Unlabeled 0.5mL Each for $321.00

NBP2328451M

 
novus biologicals canada
NBP2328451ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone SFL23.6
Host Species Mouse
Isotype κ
Immunogen Hepatocytes from 20-22 week-old human fetus
Antigen Erythrocyte Specific
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL