Please login to your online account to display your discounted pricing

anti-Erythrocyte Specific, Clone: SFL23.6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328451ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328451M

  • / Each
 

Description & Specifications

Specifications

Antigen Erythrocyte Specific
Quantity 1mL
Regulatory Status RUO
Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen Hepatocytes from 20-22 week-old human fetus
Isotype κ
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Clone SFL23.6
Host Species Mouse