Please login to your online account to display your discounted pricing

anti-ERManI, Clone: 3C2, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 3C2
Host Species Murine
Isotype κ
Immunogen Full length human recombinant ERManI [Swiss-Prot No. Q9UKM7]
Antigen ERManI
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP213167SS

 
novus biologicals canada
NBP213167SS
Unlabeled 0.025mL Each for $109.00

NBP213167

 
novus biologicals canada
NBP213167
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 3C2
Host Species Murine
Isotype κ
Immunogen Full length human recombinant ERManI [Swiss-Prot No. Q9UKM7]
Antigen ERManI
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Human. Does not cross react with Mouse.
Gene Accession No. Q9UKM7
Gene ID (Entrez) 11253
Formulation Tris-glycine, 150mM NaCl pH 7.5
Concentration 1.0mg/mL