Please login to your online account to display your discounted pricing

anti-ErbB2/Her2, Clone: HRB2/718, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328641ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328641M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Recombinant human HER-2 protein
Isotype κ
Antigen ErbB2/Her2
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P04626
Concentration 0.2mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 2064
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Clone HRB2/718
Cross Reactivity Human
Quantity 1mL