Please login to your online account to display your discounted pricing

anti-ErbB2/Her2, Clone: HRB2/282, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328631ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328631M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype κ
Regulatory Status RUO
Cross Reactivity Human
Clone HRB2/282
Immunogen Recombinant human HER-2 protein
Host Species Mouse
Gene Accession No. P04626
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2064
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Concentration 0.2mg/mL
Primary or Secondary Primary
Quantity 1mL
Format Purified
Antigen ErbB2/Her2