Please login to your online account to display your discounted pricing

anti-ErbB2/Her2, Clone: HRB2/282, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232863

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232863

  • / Each
 

Description & Specifications

Specifications

Antigen ErbB2/Her2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone HRB2/282
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04626
Host Species Mouse
Immunogen Recombinant human HER-2 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2064