Please login to your online account to display your discounted pricing

anti-ErbB2/Her2, Clone: HRB2/282, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HRB2/282
Host Species Mouse
Isotype κ
Immunogen Recombinant human HER-2 protein
Antigen ErbB2/Her2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232863

 
novus biologicals canada
NBP232863
Unlabeled 0.5mL Each for $321.00

NBP2328631M

 
novus biologicals canada
NBP2328631ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone HRB2/282
Host Species Mouse
Isotype κ
Immunogen Recombinant human HER-2 protein
Antigen ErbB2/Her2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P04626
Gene ID (Entrez) 2064
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL