Please login to your online account to display your discounted pricing

anti-ErbB2/Her2, Clone: HRB2/258, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328621ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328621M


Description & Specifications


Antigen ErbB2/Her2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone HRB2/258
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04626
Host Species Mouse
Immunogen Recombinant human HER-2 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2064