Please login to your online account to display your discounted pricing

anti-EPX, Clone: EPO104, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232844

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232844

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Isotype κ
Antigen EPX
Quantity 0.5mL
Regulatory Status RUO
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Gene Accession No. P11678
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8288
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Primary or Secondary Primary
Clone EPO104
Host Species Mouse