Please login to your online account to display your discounted pricing

anti-EPX, Clone: EPO104, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone EPO104
Host Species Mouse
Isotype κ
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Antigen EPX
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232844

 
novus biologicals canada
NBP232844
Unlabeled 0.5mL Each for $321.00

NBP2328441M

 
novus biologicals canada
NBP2328441ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone EPO104
Host Species Mouse
Isotype κ
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Antigen EPX
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Gene Accession No. P11678
Gene ID (Entrez) 8288
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL