Please login to your online account to display your discounted pricing

anti-EPX, Clone: AHE-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232964

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232964

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Quantity 0.5mL
Antigen EPX
Host Species Mouse
Cross Reactivity Human
Gene Accession No. P11678
Clone AHE-1
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8288
Primary or Secondary Primary
Conjugate Unlabeled
Isotype κ
Concentration 0.2mg/mL
Regulatory Status RUO