Please login to your online account to display your discounted pricing

anti-EPX, Clone: AHE-1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2329641ML

 View all options for this product


For Research Use Only

Catalog No. NBP2329641M


Description & Specifications


Antigen EPX
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone AHE-1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P11678
Host Species Mouse
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8288