Please login to your online account to display your discounted pricing

anti-Eps8, Clone: 18F1F2, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 18F1F2
Host Species Murine
Immunogen Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody.
Antigen Eps8
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227300

 
novus biologicals canada
NBP227300
Unlabeled 0.1mg Each for $259.02

NBP227300SS

 
novus biologicals canada
NBP227300SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 18F1F2
Host Species Murine
Immunogen Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody.
Antigen Eps8
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q12929
Gene ID (Entrez) 2059
Formulation 0.1mg EPS8 antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL