Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Clone: GZ-20, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2223891ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2223891M

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Clone GZ-20
Cross Reactivity Rat
Antigen EpCAM/TROP1
Immunogen Rat small intestine cells were used as the immunogen for the EpCAM antibody.
Host Species Mouse
Primary or Secondary Primary
Formulation Culture Medium
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Epitope of GZ-1 and GZ-20 monoclonal antibodies are different. EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. Human (Negative).
Conjugate Unlabeled
Quantity 1mL
Regulatory Status RUO