Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Clone: GZ-20, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone GZ-20
Host Species Mouse
Isotype IgG1
Immunogen Rat small intestine cells were used as the immunogen for the EpCAM antibody.
Antigen EpCAM/TROP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222389

 
novus biologicals canada
NBP222389
Unlabeled 0.5mL Each for $321.00

NBP2223891M

 
novus biologicals canada
NBP2223891ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone GZ-20
Host Species Mouse
Isotype IgG1
Immunogen Rat small intestine cells were used as the immunogen for the EpCAM antibody.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Epitope of GZ-1 and GZ-20 monoclonal antibodies are different. EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. Human (Negative).
Gene ID (Entrez) 4072
Formulation Culture Medium