Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Clone: GZ-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2223871ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2223871M

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1mL
Cross Reactivity Rat
Antigen EpCAM/TROP1
Format Concentrated
Immunogen Rat small intestine cells were used as the immunogen for the EpCAM antibody.
Primary or Secondary Primary
Clone GZ-1
Monoclonal or Polyclonal Monoclonal
Test Specificity Epitope of GZ-1 and GZ-20 monoclonal antibodies are different. EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas.
Conjugate Unlabeled
Formulation Culture Medium
Isotype IgG1
Gene ID (Entrez) 4072
Regulatory Status RUO
Host Species Mouse