Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Clone: AUA1, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone AUA1
Host Species Murine
Isotype IgG1
Immunogen LoVo cell line preparation (Human).
Antigen EpCAM/TROP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001182SS

 
novus biologicals canada
NB6001182SS
Unlabeled 0.025mL Each for $139.00

NB6001182

 
novus biologicals canada
NB6001182
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone AUA1
Host Species Murine
Isotype IgG1
Immunogen LoVo cell line preparation (Human).
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Cross-reacts with Human. Not yet tested in other species.
Gene ID (Entrez) 4072
Formulation PBS
Concentration 1.0mg/mL