Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Clone: 60N5D8, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 60N5D8
Host Species Murine
Isotype κ
Immunogen Within the range of amino acids 289-335 were used as the immunogen for the EpCAM antibody.
Antigen EpCAM/TROP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222381

 
novus biologicals canada
NBP222381
Unlabeled 0.1mg Each for $309.05

NBP222381SS

 
novus biologicals canada
NBP222381SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 60N5D8
Host Species Murine
Isotype κ
Immunogen Within the range of amino acids 289-335 were used as the immunogen for the EpCAM antibody.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. NP_612550.1
Gene ID (Entrez) 4072
Formulation 0.1mg antibody in 0.2mL in PBS and 0.05% BSA
Concentration 0.5mg/mL